missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxysalicylic acid, 99%

Product Code. 10592961 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10592961 50 g Glass bottle 50g
10562771 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10592961 Supplier Thermo Scientific Acros Supplier No. 174240500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2237-36-7 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2237-36-7
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.15
MDL Number MFCD00002450
InChI Key MRIXVKKOHPQOFK-UHFFFAOYSA-N
Synonym 4-methoxysalicylic acid, 2-hydroxy-p-anisic acid, 2-hydroxy-4-methoxy-benzoic acid, benzoic acid, 2-hydroxy-4-methoxy, 2-hydroxy-4-methoxybenzoes, 4-methoxysalicylicacid, pubchem18895, 4-methoxy salicylic acid, 4-methoxy-salicylic acid, acmc-1cs6b
PubChem CID 75231
IUPAC Name 2-hydroxy-4-methoxybenzoic acid
SMILES COC1=CC=C(C(O)=O)C(O)=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 157.0°C to 159.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3OC6H3(OH)CO2H
Quantity 50 g
Beilstein 10, 379
Formula Weight 168.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methoxysalicylic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.