missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxystyrene, 96%, stabilized

Product Code. 10675022 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10675022 25 g Glass bottle 25g
10531991 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10675022 Supplier Thermo Scientific Acros Supplier No. 161390250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 637-69-4 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-69-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008619
InChI Key UAJRSHJHFRVGMG-UHFFFAOYSA-N
Synonym 4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
PubChem CID 12507
IUPAC Name 1-ethenyl-4-methoxybenzene
SMILES COC1=CC=C(C=C)C=C1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 204°C to 205°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC6H4OCH3
Refractive Index 1.5600 to 1.562
Quantity 25 g
Beilstein 06, 561
Specific Gravity 1
Stabilizer 0.1% 4-tert-butylcatechol
Formula Weight 134.18
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 4-Methoxystyrene, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.