missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxystyrene, 98%, stab. with 0.1% 4-tert-butylcatechol

Product Code. 11436076
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436076 25 g 25g
11426076 5 g 5g
11446076 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11436076 Supplier Thermo Scientific Alfa Aesar Supplier No. B22353.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 637-69-4 | C9H10O | 134.18 g/mol

4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxystyrene is employed in the ferric chloride-catalyzed addition of activated methylenes to styrenes. It acts as a monomer in polymerization reactions. Further, it is used to prepare 1,1,2,2-Tetracyano-3-(p-methoxyphenyl)cyclobutane by reacting with ethenetetracarbonitrile.

Solubility
Slightly miscible with methanol.

Notes
Light sensitive. Store in cool place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-69-4
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008619
InChI Key UAJRSHJHFRVGMG-UHFFFAOYSA-N
Synonym 4-methoxystyrene, 1-methoxy-4-vinylbenzene, p-methoxystyrene, 4-vinylanisole, benzene, 1-ethenyl-4-methoxy, p-vinylanisole, anisole, p-vinyl, unii-2ish8t4a6e, ccris 4381, 4-vinylanisol
PubChem CID 12507
IUPAC Name 1-ethenyl-4-methoxybenzene
SMILES COC1=CC=C(C=C)C=C1

Specifications

Density 1.002
Boiling Point 204°C to 205°C
Flash Point 72°C (161°F)
Refractive Index 1.561
Quantity 25 g
Beilstein 1098935
Solubility Information Slightly miscible with methanol.
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material 4-Methoxystyrene, stabilized with 0.1% 4-tert-butylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.