missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxytrityl chloride, 97%

Product Code. 11488823
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488823 250 g 250g
11468823 10 g 10g
11478823 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488823 Supplier Thermo Scientific Alfa Aesar Supplier No. A10545.30
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14470-28-1 | C20H17ClO | 308.805 g/mol

It is used as selective protecting reagent that is used for primary hydroxyl groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as selective protecting reagent that is used for primary hydroxyl groups.

Solubility
Soluble in toluene.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14470-28-1
Molecular Formula C20H17ClO
Molecular Weight (g/mol) 308.805
MDL Number MFCD00000814
InChI Key OBOHMJWDFPBPKD-UHFFFAOYSA-N
Synonym 4-methoxytriphenylchloromethane, 4-methoxytrityl chloride, 4-methoxytriphenylmethyl chloride, 4-monomethoxytrityl chloride, mmtrcl, p-chlorodiphenylmethyl anisole, p-anisylchlorodiphenylmethane, 1-chlorodiphenylmethyl-4-methoxybenzene, chloro 4-methoxyphenyl methylene dibenzene, 4-methoxytritylchloride
PubChem CID 84462
IUPAC Name 1-[chloro(diphenyl)methyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)Cl

Specifications

Melting Point 121°C to 125°C
Quantity 250 g
Beilstein 798425
Sensitivity Moisture sensitive
Solubility Information Soluble in toluene.
Formula Weight 308.81
Percent Purity 97%
Chemical Name or Material 4-Methoxytrityl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.