missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-oxazole-5-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10620535
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10620535 250 mg 250mg
10565073 1 g 1g
10451284 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10620535 Supplier Thermo Scientific Maybridge Supplier No. CC10001CB
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 2510-32-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.099
InChI Key ZIXUNDOOBLSXPE-UHFFFAOYSA-N
Synonym 4-methyloxazole-5-carboxylic acid, 4-methyl-5-oxazolecarboxylic acid, 5-oxazolecarboxylic acid, 4-methyl, 4-methyl-oxazole-5-carboxylic acid, ksc205g3h, 4-methyl-5-oxazolecarboxylicacid, 4-methyl-oxazol-5-carboxylic acid, 4-methoxyoxazole-5-carboxylic acid, 5-oxazolecarboxylicacid, 4-methyl
PubChem CID 292311
IUPAC Name 4-methyl-1,3-oxazole-5-carboxylic acid
SMILES CC1=C(OC=N1)C(=O)O

Specifications

Assay Percent Range 95%
Quantity 250 mg
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 4-Methyl-1, 3-oxazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.