missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10643233
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10643233 1 g Amber Glass Bottle 1g
10522473 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10643233 Supplier Thermo Scientific Maybridge Supplier No. SPB03027DA
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 144059-86-9
Molecular Formula C12H8F3NO2S
Molecular Weight (g/mol) 287.256
MDL Number MFCD00068105
InChI Key DRFFZMPSUPHSJN-UHFFFAOYSA-N
Synonym 4-methyl-2-4-trifluoromethyl phenyl thiazole-5-carboxylic acid, 4-methyl-2-4-trifluoromethyl phenyl-1,3-thiazole-5-carboxylic acid, 4-methyl-2-4-trifluoromethylphenyl thiazole-5-carboxylic acid, 2-4-trifluoromethyl phenyl-4-methylthiazole-5-carboxylic acid, 4-methyl-2-4-trifluoromethyl-phenyl-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 4-methyl-2-4-trifluoromethyl phenyl, 5-thiazolecarboxylicacid, 4-methyl-2-4-trifluoromethyl phenyl, maybridge1_008801
PubChem CID 2775663
IUPAC Name 4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)C(F)(F)F)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 287.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-[4-(trifluoromethyl)phenyl]-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.