missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(4-(trifluoromethyl)phenyl)thiazole-5-carboxylic Acid, TRC

Product Code. 30136912
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136912 1 g 1g
30133983 500 mg 500mg
30009137 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136912 Supplier Toronto Research Chemicals Supplier No. TRCB4860701G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144059-86-9
Molecular Formula C12 H8 F3 N O2 S
Molecular Weight (g/mol) 287.26
Synonym 4-Methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolecarboxylic Acid;
IUPAC Name 4-methyl-2-[4-(trifluoromethyl)phenyl]thiazole-5-carboxylic acid
SMILES Cc1nc(sc1C(=O)O)c2ccc(cc2)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H8F3NO2S/c1-6-9(11(17)18)19-10(16-6)7-2-4-8(5-3-7)12(13,14)15/h2-5H,1H3,(H,17,18)
Formula Weight 287.0228
Chemical Name or Material 4-Methyl-2-(4-(trifluoromethyl)phenyl)thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.