missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitroaniline, 99%

Product Code. 10562331 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562331 100 g Glass bottle 100g
10616522 5 g Glass bottle 5g
10225130 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10562331 Supplier Thermo Scientific Acros Supplier No. 127241000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 89-62-3 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-62-3
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007907
InChI Key DLURHXYXQYMPLT-UHFFFAOYSA-N
Synonym 2-nitro-p-toluidine, 4-amino-3-nitrotoluene, 3-nitro-4-toluidine, fast red gl, 3-nitro-4-aminotoluene, fast red gl base, benzenamine, 4-methyl-2-nitro, 2-nitro-4-methylaniline, 4-methyl-6-nitroaniline, mnpt
PubChem CID 6978
ChEBI CHEBI:66920
IUPAC Name 4-methyl-2-nitroaniline
SMILES CC1=CC=C(N)C(=C1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 112°C to 115°C
Color Brown-Orange to Orange
Boiling Point 169°C (21.0 mmHg)
Flash Point 157°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 100 g
Beilstein 12, 1000
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol and diethyl ether
Formula Weight 152.15
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Methyl-2-nitroaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-2-nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.