missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitrophenol, 99%

Product Code. 10514061 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10514061 100 g Glass bottle 100g
10091521 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10514061 Supplier Thermo Scientific Acros Supplier No. 127311000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 119-33-5 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-33-5
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007120
InChI Key SYDNSSSQVSOXTN-UHFFFAOYSA-N
Synonym 2-nitro-p-cresol, phenol, 4-methyl-2-nitro, o-nitro-p-cresol, 2-nitro-4-methylphenol, 4-hydroxy-3-nitrotoluene, p-cresol, 2-nitro, 2-nitro-4-cresol, unii-p92kpk2nl3, 4-methyl-2-nitro-phenol, p92kpk2nl3
PubChem CID 8391
IUPAC Name 4-methyl-2-nitrophenol
SMILES CC1=CC(=C(C=C1)O)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 30.0°C to 34.0°C
Density 1.2400g/mL
Boiling Point 234.0°C
Flash Point 108°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)OH
Quantity 100 g
Beilstein 06, 412
Specific Gravity 1.24
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 153.14
Percent Purity 99%
Chemical Name or Material 4-Methyl-2-nitrophenol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methyl-2-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.