missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitrophenyl isothiocyanate, 95%, Thermo Scientific™

Product Code. 11348435
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348435 5 g 5g
11338435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348435 Supplier Thermo Scientific Alfa Aesar Supplier No. L10438.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17614-74-3
Molecular Formula C8H6N2O2S
Molecular Weight (g/mol) 194.208
MDL Number MFCD00041248
InChI Key CUQUTWSUXWDJKF-UHFFFAOYSA-N
Synonym 4-methyl-2-nitrophenyl isothiocyanate, 4-methyl-2-nitrophenylisothiocyanate, benzene,1-isothiocyanato-4-methyl-2-nitro, 4-methyl-2-nitrobenzenisothiocyanate, acmc-1brfl, 4-methy-2-nitrophenyl isothiocyanate, 1-isothiocyanato-4-methyl-2-nitro-benzene, #
PubChem CID 140258
IUPAC Name 1-isothiocyanato-4-methyl-2-nitrobenzene
SMILES CC1=CC(=C(C=C1)N=C=S)[N+](=O)[O-]

Specifications

Melting Point 62°C to 67°C
Odor Irritating
Quantity 5 g
UN Number UN2923
Beilstein 2969500
Sensitivity Moisture sensitive
Formula Weight 194.21
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-nitrophenyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.