missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(trifluoromethyl)benzonitrile, 98%

Product Code. 15474557
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15474557 5 g 5g
15464557 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474557 Supplier Thermo Scientific Alfa Aesar Supplier No. H31904.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 261952-05-0 | C9H6F3N | 185.15 g/mol

It is used as primary and secondary reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as primary and secondary reagent.

Solubility
Insoluble in water.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261952-05-0
Molecular Formula C9H6F3N
Molecular Weight (g/mol) 185.15
MDL Number MFCD01631596
InChI Key WCZWEODOIUKVHX-UHFFFAOYSA-N
Synonym 4-methyl-2-trifluoromethyl benzonitrile, 4-methyl-2-trifluoromethyl benzenecarbonitrile, 2-cyano-5-methylbenzotrifluoride, 2-trifluoromethyl-p-tolunitrile, 4-cyano-3-trifluoromethyl toluene, benzonitrile,4-methyl-2-trifluoromethyl
PubChem CID 2775597
IUPAC Name 4-methyl-2-(trifluoromethyl)benzonitrile
SMILES CC1=CC(=C(C=C1)C#N)C(F)(F)F

Specifications

Melting Point 22°C to 28°C
Quantity 5 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 185.15
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-(trifluoromethyl)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.