missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-nitroaniline, 97%

Product Code. 10222702 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10222702 5 g Glass bottle 5g
10669834 25 g Glass bottle 25g
10783954 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10222702 Supplier Thermo Scientific Acros Supplier No. 127250050
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 119-32-4 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-32-4
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007910
InChI Key GDIIPKWHAQGCJF-UHFFFAOYSA-N
Synonym 4-amino-2-nitrotoluene, 3-nitro-4-methylaniline, 3-nitro-p-toluidine, m-nitro-p-toluidine, 2-nitro-4-aminotoluene, benzenamine, 4-methyl-3-nitro, 5-nitro-4-toluidine, p-toluidine, 3-nitro, gl-amin czech, gl-amin
PubChem CID 8390
ChEBI CHEBI:81670
IUPAC Name 4-methyl-3-nitroaniline
SMILES CC1=C(C=C(C=C1)N)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 74°C to 78°C
Color Brown or Ochre to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 5 g
Beilstein 12, 996
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methyl-3-nitroaniline
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methyl-3-nitroaniline
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.