missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-phenylpentan-2-one, TRC

Product Code. 30481013
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481013 50 mg 50mg
30466768 10 mg 10mg
30470870 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481013 Supplier Toronto Research Chemicals Supplier No. TRCM35666850MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15934-57-3
Molecular Formula C12 H16 O
Molecular Weight (g/mol) 176.255
Synonym 3-Phenyl-4-methylpentan-2-one
IUPAC Name 4-methyl-3-phenylpentan-2-one
SMILES CC(C)C(C(=O)C)c1ccccc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H16O/c1-9(2)12(10(3)13)11-7-5-4-6-8-11/h4-9,12H,1-3H3
Formula Weight 176.12
Chemical Name or Material 4-Methyl-3-phenylpentan-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.