missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetanilide, 98+%

Product Code. p-7021402
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11494724

Brand: Thermo Scientific Alfa Aesar A13721.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-89-9 | C9H11NO | 149.193 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-89-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00008677
InChI Key YICAMJWHIUMFDI-UHFFFAOYSA-N
Synonym 4'-methylacetanilide, p-acetotoluidide, n-4-methylphenyl acetamide, 4-methylacetanilide, n-acetyl-p-toluidine, p-acetotoluide, acetyl-p-toluidine, p-acetotoluidine, acetamide, n-4-methylphenyl, 4-acetotoluide
PubChem CID 7684
IUPAC Name N-(4-methylphenyl)acetamide
SMILES CC1=CC=C(C=C1)NC(=O)C

Specifications

Melting Point 147°C to 150°C
Boiling Point 306°C to 308°C
Quantity 25 g
Beilstein 607036
Merck Index 14,74
Formula Weight 149.19
Percent Purity ≥98%
Chemical Name or Material 4'-Methylacetanilide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.