missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetanilide, TRC

Product Code. 30132822
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132822 25 g 25g
30135062 10 g 10g
30010644 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132822 Supplier Toronto Research Chemicals Supplier No. TRCM27501025G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-89-9
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
Synonym N-(4-methylphenyl)acetamide, 4-(Acetylamino)toluene, 4-Acetotoluide, 1-Acetamido-4-methylbenzene, N-p-Tolylacetamide, p-Acetotoluide, NSC 7644;
IUPAC Name N-(4-methylphenyl)acetamide
SMILES CC(=O)Nc1ccc(C)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H11NO/c1-7-3-5-9(6-4-7)10-8(2)11/h3-6H,1-2H3,(H,10,11)
Formula Weight 149.0841
Chemical Name or Material 4'-Methylacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.