missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetanilide, TRC

Product Code. 30010644
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010644 1 g 1g
30132822 25 g 25g
30135062 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010644 Supplier Toronto Research Chemicals Supplier No. TRCM2750101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-89-9
InChI Formula InChI=1S/C9H11NO/c1-7-3-5-9(6-4-7)10-8(2)11/h3-6H,1-2H3,(H,10,11)
IUPAC Name N-(4-methylphenyl)acetamide
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
Chemical Name or Material 4'-Methylacetanilide

Specifications

Quantity 1 g
Synonym N-(4-methylphenyl)acetamide, 4-(Acetylamino)toluene, 4-Acetotoluide, 1-Acetamido-4-methylbenzene, N-p-Tolylacetamide, p-Acetotoluide, NSC 7644;
SMILES CC(=O)Nc1ccc(C)cc1
Formula Weight 149.0841
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.