missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylamino)butanoic Acid Hydrochloride, TRC

Product Code. 30479494
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479494 10 g 10g
30468354 1 g 1g
30482197 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30479494 Supplier Toronto Research Chemicals Supplier No. TRCM33198310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6976-17-6
Molecular Formula C5H11NO2 . HCl
Molecular Weight (g/mol) 153.61
Synonym 4-(Methylamino)butanoic Acid Hydrochloride, 4-(Methylamino)butyric Acid Hydrochloride
IUPAC Name 4-(methylamino)butanoic acid;hydrochloride
SMILES Cl.CNCCCC(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H11NO2.ClH/c1-6-4-2-3-5(7)8;/h6H,2-4H2,1H3,(H,7,8);1H
Formula Weight 153.0557
Chemical Name or Material 4-(Methylamino)butanoic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.