missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzenesulfonhydrazide, 97%

Product Code. 10606172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606172 100 g Plastic bottle 100g
10605982 25 g Plastic bottle 25g
10279593 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10606172 Supplier Thermo Scientific Acros Supplier No. 157861000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1576-35-8 | C7H10N2O2S | 186.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1576-35-8
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.23
MDL Number MFCD00007588
InChI Key ICGLPKIVTVWCFT-UHFFFAOYSA-N
Synonym 4-methylbenzenesulfonhydrazide, p-toluenesulfonhydrazide, tosylhydrazine, p-toluenesulfonyl hydrazide, tosylhydrazide, p-tosylhydrazine, p-toluenesulfonylhydrazine, celogen tsh, ptsh, 4-toluenesulfonyl hydrazide
PubChem CID 15303
SMILES CC1=CC=C(C=C1)S(=O)(=O)NN

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 109.0°C to 113.0°C
Color Gray to White or White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3C6H4SO2NHNH2
Quantity 100 g
Beilstein 11, II, 66
Fieser 01,1185; 02,417; 03,293; 04,511; 05,678; 06,598; 07,375; 08,489; 09,472
Merck Index 15, 9694
Solubility Information Solubility in water: 5g/L (15°C)
IUPAC Name 4-methylbenzene-1-sulfonohydrazide
Formula Weight 186.23
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Methylbenzenesulfonhydrazide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methylbenzenesulfonhydrazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable solid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.