missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzophenone, 98%

Product Code. 11364699
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
This item is not returnable. View return policy

Product Code. 11364699

Brand: Thermo Scientific Alfa Aesar A15479.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 134-84-9 | C14H12O | 196.249 g/mol

4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzophenone is used as clear coatings for wood, plastic and metal, overprint varnish.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place. Store in a tightly closed container. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-84-9
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD00008553
InChI Key WXPWZZHELZEVPO-UHFFFAOYSA-N
Synonym 4-methylbenzophenone, phenyl p-tolyl methanone, 4-methylphenyl phenyl methanone, p-methylbenzophenone, methanone, 4-methylphenyl phenyl, p-benzoyltoluene, 4-methyl benzophenone, phenyl p-tolyl ketone, benzophenone, 4-methyl, p-benzophenone, methyl
PubChem CID 8652
IUPAC Name (4-methylphenyl)-phenylmethanone
SMILES CC1=CC=C(C=C1)C(=O)C2=CC=CC=C2

Specifications

Melting Point 53°C to 57°C
Boiling Point 326°C
Flash Point 143°C (289°F)
Quantity 50 g
Beilstein 1909310
Merck Index 14,7317
Solubility Information Insoluble in water.
Formula Weight 196.25
Percent Purity 98%
Chemical Name or Material 4-Methylbenzophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.