missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzophenone, TRC

Product Code. 30472922
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30472922 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30472922 Supplier Toronto Research Chemicals Supplier No. TRCM2887701G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 134-84-9
Molecular Formula C14 H12 O
Quantity 1 g
Synonym Benzophenone, 4-methyl- (6CI,8CI), (4-Methylphenyl)phenylmethanone, (Phenyl)(4-methylphenyl)methanone, 4-Benzoyltoluene, 4-Methylbenzophenone, 4-Methylphenyl phenyl ketone, JRCure 1024, NSC 4898, Phenyl p-tolyl ketone, Phenyl(4-tolyl)methanone, SB-PI 712, Speedcure MBP, UV-Cure PMB, p-Benzoyltoluene, p-Methylbenzophenone, p-Methylphenyl phenyl ketone, p-Tolyl phenyl ketone
InChI Formula InChI=1S/C14H12O/c1-11-7-9-13(10-8-11)14(15)12-5-3-2-4-6-12/h2-10H,1H3
SMILES Cc1ccc(cc1)C(=O)c2ccccc2
IUPAC Name (4-methylphenyl)-phenylmethanone
Molecular Weight (g/mol) 196.2445
Formula Weight 196.0888
Percent Purity >95%
Chemical Name or Material 4-Methylbenzophenone
Grade HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.