missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl alcohol, 98%

Product Code. 10677122 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10677122 25 g Glass bottle 25g
10035973 5 g Glass bottle 5g
10486800 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10677122 Supplier Thermo Scientific Acros Supplier No. 155920250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 589-18-4 | C8H10O | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-18-4
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.17
MDL Number MFCD00004664
InChI Key KMTDMTZBNYGUNX-UHFFFAOYSA-N
Synonym 4-methylbenzyl alcohol, p-tolylmethanol, p-tolylcarbinol, 4-methylphenyl methanol, 4-tolylcarbinol, p-methylbenzyl alcohol, benzenemethanol, 4-methyl, 4-hydroxymethyl toluene, benzyl alcohol, p-methyl, 4-methylbenzenemethanol
PubChem CID 11505
ChEBI CHEBI:1895
IUPAC Name (4-methylphenyl)methanol
SMILES CC1=CC=C(C=C1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 61.0°C
Color White
Boiling Point 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4CH2OH
Quantity 25 g
Beilstein 06, 498
Formula Weight 122.17
Percent Purity 98%
Physical Form Crystalline Mass, Crystals or Needles
Chemical Name or Material 4-Methylbenzyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylbenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.