missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzyl bromide, 98%

Product Code. 11420883
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420883 25 g 25g
11410883 5 g 5g
11430883 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420883 Supplier Thermo Scientific Alfa Aesar Supplier No. A13297.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 104-81-4 | C8H9Br | 185.064 g/mol

4-Methylbenzyl bromide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylbenzyl bromide is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-81-4
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000181
InChI Key WZRKSPFYXUXINF-UHFFFAOYSA-N
Synonym 4-methylbenzyl bromide, 1-bromomethyl-4-methylbenzene, p-xylyl bromide, p-methylbenzyl bromide, alpha-bromo-p-xylene, benzene, 1-bromomethyl-4-methyl, 4-bromomethyl toluene, p-bromomethyl toluene, alpha-bromo-p-xylol, 1-bromomethyl-4-methyl-benzene
PubChem CID 7721
IUPAC Name 1-(bromomethyl)-4-methylbenzene
SMILES CC1=CC=C(C=C1)CBr

Specifications

Melting Point 33°C to 36°C
Density 1.324
Boiling Point 218°C to 220°C
Flash Point 97°C (206°F)
Quantity 25 g
UN Number UN3417
Beilstein 507389
Merck Index 14,10090
Solubility Information Insoluble in water.
Formula Weight 185.07
Percent Purity 98%
Chemical Name or Material 4-Methylbenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.