missing translation for 'onlineSavingsMsg'
Learn More

4-Methylbenzylidene camphor, MedChemExpress

Product Code. 16491699
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16491699

Brand: Medchem Express HY17587

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 4-Methylbenzylidene camphor(4-MBC; Enzacamene)is an organic camphor derivative that is used in the cosmetic industry for its ability to protect the skin against UV, specifically UV B radiation.

4-Methylbenzylidene camphor(4-MBC

Specifications

Chemical Name or Material 4-Methylbenzylidene camphor
Physical Form Solid
CAS 36861-47-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C18H22O
Synonym 4-MBC Enzacamene
Solubility Information DMSO : 100 mg/mL (393.13 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC1(C)[C@@H]2CC[C@]1(C)C(/C2=C\C3=CC=C(C)C=C3)=O
Molecular Weight (g/mol) 254.37
Formula Weight 254.37
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.