missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcatechol, 98%

Product Code. 10366551 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10366551

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 452-86-8 | C7H8O2 | 124.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452-86-8
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
MDL Number MFCD00002205
InChI Key ZBCATMYQYDCTIZ-UHFFFAOYSA-N
Synonym 4-methylcatechol, 3,4-dihydroxytoluene, homocatechol, 4-methyl-1,2-benzenediol, p-methylcatechol, 4-methylpyrocatechol, homopyrocatechol, 1,2-dihydroxy-4-methylbenzene, p-methylpyrocatechol, toluene-3,4-diol
PubChem CID 9958
ChEBI CHEBI:17254
IUPAC Name 4-methylbenzene-1,2-diol
SMILES CC1=CC=C(O)C(O)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 62°C to 69°C
Color Brown to White
Boiling Point 251°C
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H3(OH)2
Quantity 100 g
Beilstein 06, 878
Solubility Information Solubility in water: soluble in water. Other solubilities: miscible with acetone,alcohol,ether and toluene,moderately soluble in petroleum ether,slightly soluble in carbon tetrachloride
Formula Weight 124.14
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-Methylcatechol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.