missing translation for 'onlineSavingsMsg'
Learn More

4-Methylcinnamic acid, predominantly trans, 99%

Product Code. 11400963
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400963 5 g 5g
11410963 25 g 25g
11420963 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400963 Supplier Thermo Scientific Alfa Aesar Supplier No. A13337.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1866-39-3 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1866-39-3
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00002697
InChI Key RURHILYUWQEGOS-VOTSOKGWSA-N
Synonym 4-methylcinnamic acid, 3-p-tolyl acrylic acid, e-3-p-tolyl acrylic acid, p-methylcinnamic acid, p-methyl cinnamic acid, 2e-3-4-methylphenyl acrylic acid, 2e-3-4-methylphenyl prop-2-enoic acid, 4-methyl-cinnamic acid, e-3-4-methylphenyl-2-propenoic acid, 3-4-methylphenyl-2-propenoic acid
PubChem CID 731767
IUPAC Name (E)-3-(4-methylphenyl)prop-2-enoic acid
SMILES CC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 196°C to 200°C
Quantity 5 g
Beilstein 1364327
Formula Weight 162.19
Percent Purity 99%
Chemical Name or Material 4-Methylcinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.