missing translation for 'onlineSavingsMsg'
Learn More

4-Methyldiphenylamine, 98%

Product Code. 11425330
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425330 10 g 10g
11435330 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425330 Supplier Thermo Scientific Alfa Aesar Supplier No. 044560.09
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 620-84-8 | C13H13N | 183.25 g/mol

4-Methyldiphenylamine is used as a syntheses material intermediates as well as OLED materials. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-84-8
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00092921
InChI Key AGHYMXKKEXDUTA-UHFFFAOYSA-N
Synonym 4-methyldiphenylamine, benzenamine, 4-methyl-n-phenyl, n-phenyl-p-toluidine, 4-methyl-n-phenyl-aniline, 4-methyl-n-phenylbenzamine, 4-methylphenyl phenylamine, phenyl-p-tolylamine, 4-methyldiphenylamin, phenyl-p-tolyl-amine, n-pnenyl-4-methylaniline
PubChem CID 12109
IUPAC Name 4-methyl-N-phenylaniline
SMILES CC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 85°C to 86°C
Color Brown
Boiling Point 334°C
Flash Point 155°C (311°F)
Quantity 10 g
Solubility Information Insoluble in water. Soluble in benzene,toluene,methanol,ethanol
Formula Weight 183.25
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 4-Methyldiphenylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.