missing translation for 'onlineSavingsMsg'
Learn More

4-Methylhippuric acid, 98%, Thermo Scientific™

Product Code. 15457248
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457248 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15457248 Supplier Thermo Scientific Alfa Aesar Supplier No. H60754.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 27115-50-0
Melting Point 163°C to 165°C
Molecular Formula C10H11NO3
MDL Number MFCD00020449
Quantity 1 g
Synonym 4-methylhippuric acid, n-4-methylbenzoyl glycine, p-toluric acid, p-methylhippuric acid, n-p-toluoyl glycine, glycine, n-4-methylbenzoyl, 4-methyl hippuric acid, 2-4-methylbenzamido acetic acid, unii-9qox0dsk6f, hippuric acid, p-methyl
InChI Key NRSCPTLHWVWLLH-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)C(=O)NCC(=O)O
IUPAC Name 2-[(4-methylbenzoyl)amino]acetic acid
Molecular Weight (g/mol) 193.202
PubChem CID 97479
ChEBI CHEBI:68552
Formula Weight 193.2
Percent Purity 98%
Chemical Name or Material 4-Methylhippuric acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.