missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylmercapto)phenol, 97%

Product Code. 11915731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11915731 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11915731 Supplier Thermo Scientific Acros Supplier No. 127130250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1073-72-9 | C7H8OS | 140.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1073-72-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 85.0°C to 87.0°C
Color Brown to White
Boiling Point 153.0°C to 156.0°C (20.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C7H8OS
Linear Formula CH3SC6H4OH
MDL Number MFCD00002351
Quantity 25 g
Beilstein 06,I,419
Fieser 02,281
Synonym 4-methylthio phenol, 4-methylmercapto phenol, phenol, 4-methylthio, p-methylthio phenol, 4-hydroxythioanisole, 4-methylmercaptophenol, p-hydroxythioanisole, 4-methylsulfanyl phenol, phenol, p-methylthio, p-hydroxyphenyl methyl sulfide
InChI Key QASBCTGZKABPKX-UHFFFAOYSA-N
SMILES CSC1=CC=C(O)C=C1
IUPAC Name 4-methylsulfanylphenol
Molecular Weight (g/mol) 140.20
PubChem CID 14086
ChEBI CHEBI:38862
Formula Weight 140.2
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-(Methylmercapto)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 4-(Methylmercapto)phenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.