missing translation for 'onlineSavingsMsg'
Learn More

4-Methylmorpholine N-oxide, 50% w/w aq. soln.

Product Code. p-7024684
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458730 100 g 100g
11408667 250 g 250g
11498657 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458730 Supplier Thermo Scientific Alfa Aesar Supplier No. A19802.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7529-22-8 | C5H11NO2 | 117.15 g/mol

4-Methylmorpholine N-oxide acts as a non-metallic catalyst for the cyanosilylation of ketones. It is also employed as a co-oxidant for Sharpless asymmetric dihydroxylation in ionic liquids. It serves as a solvent in the Lyocell process to produce tencel fiber. Further, it is used in the preparation of aldehydes from primary alcohols in the presence of tetrapropylammonium perruthenate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylmorpholine N-oxide acts as a non-metallic catalyst for the cyanosilylation of ketones. It is also employed as a co-oxidant for Sharpless asymmetric dihydroxylation in ionic liquids. It serves as a solvent in the Lyocell process to produce tencel fiber. Further, it is used in the preparation of aldehydes from primary alcohols in the presence of tetrapropylammonium perruthenate.

Solubility
Soluble in water, methanol, ethanol, acetone, ethers and dimethylsulfoxide.

Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents, acid chlorides, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7529-22-8
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00005947
InChI Key LFTLOKWAGJYHHR-UHFFFAOYSA-N
Synonym 4-methylmorpholine n-oxide, 4-methylmorpholine 4-oxide, n-methylmorpholine oxide, n-methylmorpholine n-oxide, morpholine, 4-methyl-, 4-oxide, 4-methylmorpholine-4-oxide, methyl morpholine oxide, n-methylmorpholine-n-oxide, nmo, unii-arc64pkj0f
PubChem CID 82029
ChEBI CHEBI:52093
SMILES C[N+]1([O-])CCOCC1

Specifications

Melting Point -20°C
Density 1.14
Boiling Point 118°C to 119°C
Assay Percent Range 0.5
Refractive Index 1.422
Quantity 100 g
Solubility Information Soluble in water,methanol,ethanol,acetone,ethers and dimethylsulfoxide.
IUPAC Name 4-methylmorpholin-4-ium-4-olate
Formula Weight 117.15
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Physical Form Liquid
Chemical Name or Material 4-Methylmorpholine N-oxide, w/w aq. soln.
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylmorpholine N-oxide, 50% w/w aqueous solution
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 4-Methylmorpholine N-oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.