missing translation for 'onlineSavingsMsg'
Learn More

4-Methyloxazole-5-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10103752
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10103752 1 g Amber Glass Bottle 1g
10085922 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10103752 Supplier Thermo Scientific Maybridge Supplier No. CC10002DA
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 62348-24-7
Molecular Formula C5H4ClNO2
Molecular Weight (g/mol) 145.54
MDL Number MFCD06200856
InChI Key YPKNOSGIABPXKS-UHFFFAOYSA-N
Synonym 4-methyloxazole-5-carbonyl chloride, 5-oxazolecarbonyl chloride, 4-methyl, 4-methyloxazole-5-carbonylchloride, 4-methyl-5-oxazolecarbonyl chloride, 4-methyl oxazole-5-carbonyl chloride, 4-methyl-oxazole-5-carbonyl chloride, 4-methyl-oxazole-5-carbonyl-chloride, 4-methyloxazole-5-carboxylic acid chloride, 5-oxazolecarbonyl chloride, 4-methyl-9ci
PubChem CID 2795217
IUPAC Name 4-methyl-1,3-oxazole-5-carbonyl chloride
SMILES CC1=C(OC=N1)C(Cl)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 145.54
Percent Purity 97%
Chemical Name or Material 4-Methyloxazole-5-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.