missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpiperidine, 99%

Product Code. 10123180 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10123180 100 g 100g
10487130 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123180 Supplier Thermo Scientific Acros Supplier No. 127511000
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 626-58-4 | C6H13N | 99.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-58-4
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
MDL Number MFCD00006005
InChI Key UZOFELREXGAFOI-UHFFFAOYSA-N
Synonym 4-pipecoline, piperidine, 4-methyl, gamma-pipecoline, 4-methylpiperidin, .gamma.-pipecoline, 4-methyl-piperidine, 4-methylpiperdine, 4-methylpyperidine, 4-methyl piperidine, hexahydro-g-picoline
PubChem CID 69381
IUPAC Name 4-methylpiperidine
SMILES CC1CCNCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 4.0°C to 5.0°C
Color Yellow
Density 0.8400g/mL
Boiling Point 122.0°C
Flash Point 13°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4448 to 1.4468
Quantity 100 g
Specific Gravity 0.84
Solubility Information Solubility in water: miscible.
Formula Weight 99.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Methylpiperidine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methylpiperidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Flammable liquid Category 2
Hazard Statement
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.