missing translation for 'onlineSavingsMsg'
Learn More

4-Methylpyrogallol, TRC

Product Code. 17900365
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17900365 1 g 1g
17964555 5 g 5g
17971535 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17900365 Supplier Toronto Research Chemicals Supplier No. TRCM2277051G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3955-29-1
Molecular Formula C7 H8 O3
Molecular Weight (g/mol) 140.137
Synonym 4-Methyl-1,2,3-benzenetriol
IUPAC Name 4-methylbenzene-1,2,3-triol
SMILES Cc1ccc(O)c(O)c1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H8O3/c1-4-2-3-5(8)7(10)6(4)9/h2-3,8-10H,1H3
Formula Weight 140.0473
Percent Purity >95%
Chemical Name or Material 4-Methylpyrogallol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.