missing translation for 'onlineSavingsMsg'
Learn More

4-Methylstyrene, 98%, stab. with 0.1% 3,5-di-tert-butylcatechol

Product Code. 11301297
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11301297

Brand: Thermo Scientific Alfa Aesar L01480.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 622-97-9 | C9H10 | 118.18 g/mol

4-Methylstyrene is used as a monomer for polyesters and in plastics production. It is also used as an intermediate in paint and coating additives. Further, it is used with other vinyltoluene isomers (3-vinyltoluene) as monomers for the preparation of poly(vinyltoluene). In addition to this, it is employed as a bi- ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-pi complexes. Further, it is involved in Heck coupling reactions with chlorobenzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methylstyrene is used as a monomer for polyesters and in plastics production. It is also used as an intermediate in paint and coating additives. Further, it is used with other vinyltoluene isomers (3-vinyltoluene) as monomers for the preparation of poly(vinyltoluene). In addition to this, it is employed as a bi- ligand in the preparation of cationic, two-coordinate triphenylphosphine-gold(I)-pi complexes. Further, it is involved in Heck coupling reactions with chlorobenzene.

Solubility
Miscible with benzene. slightly miscible with water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-97-9
Molecular Formula C9H10
Molecular Weight (g/mol) 118.18
MDL Number MFCD00008621
InChI Key JLBJTVDPSNHSKJ-UHFFFAOYSA-N
Synonym 4-methylstyrene, 1-methyl-4-vinylbenzene, p-methylstyrene, 4-vinyltoluene, p-vinyltoluene, benzene, 1-ethenyl-4-methyl, styrene, p-methyl, p-methyl styrene, 1-p-tolylethene, para-methylstyrene
PubChem CID 12161
IUPAC Name 1-ethenyl-4-methylbenzene
SMILES CC1=CC=C(C=C)C=C1

Specifications

Melting Point -34°C
Density 0.896
Boiling Point 171°C to 175°C
Flash Point 45°C (113°F)
Refractive Index 1.543
Quantity 10 g
UN Number UN2618
Beilstein 1209317
Solubility Information Miscible with benzene. slightly miscible with water.
Formula Weight 118.18
Percent Purity ≥98%
Chemical Name or Material 4-Methylstyrene, stabilized with 0.1% 3,5-di-tert-butylcatechol
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4-Methylstyrene
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 1B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H340-May cause genetic defects.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Methylstyrene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.