missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfanyl)phenylboronic acid, 95%, May contain varying amounts of anhydride, Maybridge

Product Code. 10525783
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Product Code. Quantity Packaging unitSize
10525783 1 g Amber Glass Bottle 1g
1 options
This item is not returnable. View return policy

Product Code. 10525783

Brand: Thermo Scientific Maybridge AC34468DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 98546-51-1
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H9BO2S
Linear Formula CH3SC6H4B(OH)2
MDL Number MFCD00093410
Quantity 1 g
Synonym 4-methylthio phenylboronic acid, 4-thioanisoleboronic acid, 4-methylsulfanyl phenylboronic acid, 4-methylthiophenylboronic acid, 4-methylthio phenyl boronic acid, 4-methylsulfanylphenyl boronic acid, 4-methylthio phenylboronicacid, 4-methylthio benzeneboronic acid, thioanisole-4-boronic acid, 4-methylmercapto benzeneboronic acid
InChI Key IVUHTLFKBDDICS-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)SC)(O)O
IUPAC Name (4-methylsulfanylphenyl)boronic acid
Molecular Weight (g/mol) 168.017
PubChem CID 2733959
Formula Weight 168.02
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfanyl)phenylboronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.