missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzeneboronic acid, 98%

Product Code. 11348656
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11348656

Brand: Thermo Scientific Alfa Aesar L17720.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 149104-88-1 | C7H9BO4S | 200.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149104-88-1
Molecular Formula C7H9BO4S
Molecular Weight (g/mol) 200.02
MDL Number MFCD01630820
InChI Key VDUKDQTYMWUSAC-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenylboronic acid, 4-methanesulfonyl phenylboronic acid, 4-methylsulfonyl phenyl boronic acid, 4-methanesulfonyl benzeneboronic acid, 4-methanesulfonylphenylboronic acid, 4-methylsulfonylphenyl boronic acid, 4-boronophenyl methyl sulphone, 4-methylsulphonyl phenylboronic acid, 4-methylsulphonyl benzeneboronic acid, 4-methanesulfonylphenyl boranediol
PubChem CID 2734364
IUPAC Name (4-methylsulfonylphenyl)boronic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)B(O)O

Specifications

Melting Point ∼200°C
Quantity 250 mg
Formula Weight 200.02
Percent Purity 98%
Chemical Name or Material 4-(Methylsulfonyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.