missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11459360
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11459360

Brand: Thermo Scientific Alfa Aesar H50241.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53606-06-7
Molecular Formula C8H9BrO2S
Molecular Weight (g/mol) 249.122
MDL Number MFCD00185849
InChI Key HGKPAXHJTMHWAH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix
PubChem CID 2733581
IUPAC Name 1-(bromomethyl)-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)CBr

Specifications

Quantity 250 mg
UN Number UN3261
Formula Weight 249.13
Percent Purity 97%
Chemical Name or Material 4-(Methylsulfonyl)benzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.