missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11469360
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469360 1 g 1g
11459360 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469360 Supplier Thermo Scientific Alfa Aesar Supplier No. H50241.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53606-06-7
Molecular Formula C8H9BrO2S
Molecular Weight (g/mol) 249.122
MDL Number MFCD00185849
InChI Key HGKPAXHJTMHWAH-UHFFFAOYSA-N
Synonym 1-bromomethyl-4-methylsulfonyl benzene, 4-methylsulfonyl benzyl bromide, 1-bromomethyl-4-methanesulfonylbenzene, benzene, 1-bromomethyl-4-methylsulfonyl, 4-methylsulphonylbenzyl bromide, 4-methylsulfonyl benzylbromide, 1-bromomethyl-4-methanesulfonyl-benzene, 1-bromomethyl-4-methylsulfonylbenzene, pubchem18666, acmc-1b0ix
PubChem CID 2733581
IUPAC Name 1-(bromomethyl)-4-methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=C(C=C1)CBr

Specifications

Quantity 1 g
UN Number UN3261
Formula Weight 249.13
Percent Purity 97%
Chemical Name or Material 4-(Methylsulfonyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.