missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenylacetic acid, 97%

Product Code. 11318866
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318866 5 g 5g
11328866 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318866 Supplier Thermo Scientific Alfa Aesar Supplier No. L19504.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90536-66-6 | C9H10O4S | 214.235 g/mol

4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90536-66-6
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00216495
InChI Key HGGWOSYNRVOQJH-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenylacetic acid, 4-methylsulphonylphenylacetic acid, 2-4-methylsulfonyl phenyl acetic acid, 4-mesylphenylacetic acid, 4-methanesulfonylphenyl acetic acid, 4-methylsulfonyl phenyl acetic acid, 4-methanesulfonyl phenylacetic acid, 2-4-methanesulfonylphenyl acetic acid, 4-methylsulfonylphenylacetic acid
PubChem CID 572345
IUPAC Name 2-(4-methylsulfonylphenyl)acetic acid
SMILES CS(=O)(=O)C1=CC=C(C=C1)CC(=O)O

Specifications

Melting Point 134°C to 138°C
Quantity 5 g
Beilstein 2646649
Solubility Information Slightly soluble in water.
Formula Weight 214.24
Percent Purity 97%
Chemical Name or Material 4-(Methylsulfonyl)phenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.