missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenylacetic Acid, TRC

Product Code. 30480469
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480469 10 g 10g
30470197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30480469 Supplier Toronto Research Chemicals Supplier No. TRCM32725510G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90536-66-6
Molecular Formula C9 H10 O4 S
Molecular Weight (g/mol) 214.24
Synonym Benzeneacetic acid, 4-(methylsulfonyl)-, Acetic acid, [p-(methylsulfonyl)phenyl]- (7CI), (4-Methanesulfonylphenyl)acetic acid, 2-[4-(Methylsulfonyl)phenyl]acetic acid, 4-(Methylsulfonyl)benzeneacetic acid, 4-(Methylsulfonyl)phenylacetic acid, 4-Mesylphenylacetic acid
IUPAC Name 2-(4-methylsulfonylphenyl)acetic acid
SMILES CS(=O)(=O)c1ccc(CC(=O)O)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H10O4S/c1-14(12,13)8-4-2-7(3-5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)
Formula Weight 214.03
Chemical Name or Material 4-(Methylsulfonyl)phenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.