missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylsulfonyl)phenylhydrazine Hydrochloride, 95%

Product Code. 10562754 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10562754 10 g Glass bottle 10g
10230001 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10562754 Supplier Thermo Scientific Acros Supplier No. 424960100
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17852-67-4 | C7H10N2O2S·HCl | 222.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17852-67-4
Molecular Formula C7H10N2O2S·HCl
Molecular Weight (g/mol) 222.69
MDL Number MFCD00216494
InChI Key QVCMFSJTNVQJFG-UHFFFAOYSA-N
Synonym 4-methylsulfonyl phenyl hydrazine hydrochloride, 4-methylsulfonyl phenylhydrazine hydrochloride, 4-methylsulphonyl phenylhydrazine hydrochloride, 4-methylsulphonylphenylhydrazine hydrochloride, 4-methanesulfonylphenyl hydrazine hydrochloride, 4-methylsulfonylphenylhydrazine hydrochloride, hydrazine, 4-methylsulfonyl phenyl-, monohydrochloride, 4-methylsulfonyl phenylhydrazine, chloride, 4-methylsulfonyl phenylhydrazine hydrochloric acid
PubChem CID 2735181
IUPAC Name (4-methylsulfonylphenyl)hydrazine;hydrochloride
SMILES CS(=O)(=O)C1=CC=C(C=C1)NN.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 202°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula H2NHNC6H4SO2CH3·HCl
Quantity 10 g
Solubility Information Solubility in water: soluble
Formula Weight 222.69
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylsulfonyl)phenylhydrazine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.