missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonyl benzaldehyde, 95%

Product Code. 10517382 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10517382 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10517382 Supplier Thermo Scientific Acros Supplier No. 424900025
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5398-77-6 | C8H8O3S | 184.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5398-77-6
CAS Min % 94.0
CAS Max % 100.0
Melting Point 155°C to 161°C
Color Beige-Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8O3S
MDL Number MFCD00216485
Quantity 2.5 g
Synonym 4-methylsulfonyl benzaldehyde, 4-methylsulphonyl benzaldehyde, 4-methanesulfonylbenzaldehyde, 4-methanesulfonyl-benzaldehyde, 4-methylsulfonyl-benzaldehyde, 4-methylsulphonylbenzaldehyde, p-methylsulphonyl benzaldehyde, benzaldehyde, 4-methylsulfonyl, pubchem8151
Solubility Information Solubility in water: insoluble
InChI Key PSVPUHBSBYJSMQ-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(C=C1)C=O
IUPAC Name 4-methylsulfonylbenzaldehyde
Molecular Weight (g/mol) 184.22
PubChem CID 220376
Formula Weight 184.22
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 4-Methylsulphonyl benzaldehyde, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylsulphonyl benzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.