missing translation for 'onlineSavingsMsg'
Learn More

4-Methylsulphonyl benzonitrile, 95%, Thermo Scientific™

Product Code. 10321221 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
Packaging:
Glass bottle
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10321221 2.5 g Glass bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10321221 Supplier Thermo Scientific Acros Supplier No. 424890025
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 22821-76-7
CAS Min % 94.0
CAS Max % 100.0
Melting Point 137.0°C to 142.0°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C8H7NO2S
MDL Number MFCD00216489
Quantity 2.5 g
Synonym 4-methylsulfonyl benzonitrile, 4-methanesulfonyl-benzonitrile, 4-methanesulfonylbenzonitrile, benzonitrile, 4-methylsulfonyl, benzonitrile, p-methylsulfonyl, 4-methylsulfonyl benzenecarbonitrile, 4-mesylbenzonitrile, maybridge3_003168, p-methylsulfonylbenzonitrile, 4-methylsulphonylbenzonitrile
Solubility Information Solubility in water: negligible
InChI Key FARXIDYHJAANGP-UHFFFAOYSA-N
SMILES CS(=O)(=O)C1=CC=C(C=C1)C#N
IUPAC Name 4-methylsulfonylbenzonitrile
Molecular Weight (g/mol) 181.21
PubChem CID 519981
Formula Weight 181.21
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylsulphonyl benzonitrile, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methylsulphonyl benzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.