missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)aniline, 98%

Product Code. 11341777
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11341777 5 g 5g
11351777 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11341777

Brand: Thermo Scientific Alfa Aesar L04950.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-96-1 | C7H9NS | 139.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-96-1
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.22
MDL Number MFCD00007889
InChI Key YKFROQCFVXOUPW-UHFFFAOYSA-N
Synonym 4-methylthio aniline, 4-methylmercapto aniline, 4-aminothioanisole, p-thioanisidine, benzenamine, 4-methylthio, p-methylthio aniline, p-aminothioanisole, p-thiomethoxyaniline, 4-methylsulfanyl aniline, 4-methylthio benzenamine
PubChem CID 66038
IUPAC Name 4-methylsulfanylaniline
SMILES CSC1=CC=C(N)C=C1

Specifications

Density 1.12
Boiling Point 272°C to 273°C
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.639
Quantity 5 g
UN Number UN2810
Beilstein 774506
Sensitivity Light sensitive
Formula Weight 139.23
Percent Purity 98%
Chemical Name or Material 4-(Methylthio)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.