missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzaldehyde, 97%

Product Code. 11497106
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11497106

Brand: Thermo Scientific Alfa Aesar B25427.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 3446-89-7 | C8H8OS | 152.211 g/mol

4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.

Solubility
Not miscible in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 3446-89-7
Molecular Formula C8H8OS
Molecular Weight (g/mol) 152.211
MDL Number MFCD00006948
InChI Key QRVYABWJVXXOTN-UHFFFAOYSA-N
Synonym 4-methylthio benzaldehyde, benzaldehyde, 4-methylthio, 4-methylsulfanyl benzaldehyde, 4-methylmercaptobenzaldehyde, p-methylthio benzaldehyde, p-methylthiobenzaldehyde, unii-1tvq11biy8, 4-aldehyde thioanisole, p-methylmercaptobenzaldehyde
PubChem CID 76985
IUPAC Name 4-methylsulfanylbenzaldehyde
SMILES CSC1=CC=C(C=C1)C=O

Spécification

Melting Point 6°C
Density 1.144
Boiling Point 118°C to 121°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.649
Quantity 25 g
Beilstein 636158
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 152.22
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzaldehyde
Afficher plus Afficher moins

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis