missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)benzoic acid, 97%

Product Code. 11431737
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431737 1 g 1g
11441737 5 g 5g
11451737 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11431737 Supplier Thermo Scientific Alfa Aesar Supplier No. B25706.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13205-48-6 | C8H8O2S | 168.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13205-48-6
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00002555
InChI Key KWHCPERWLHBLOT-UHFFFAOYSA-N
Synonym 4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa
PubChem CID 83230
IUPAC Name 4-methylsulfanylbenzoic acid
SMILES CSC1=CC=C(C=C1)C(O)=O

Specifications

Melting Point 193°C to 196°C
Quantity 1 g
Beilstein 2082809
Formula Weight 168.21
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.