missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)-m-cresol, 97%

Product Code. 11301877
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301877 5 g 5g
11311877 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301877 Supplier Thermo Scientific Alfa Aesar Supplier No. L07353.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3120-74-9 | C8H10OS | 154.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3120-74-9
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.227
MDL Number MFCD00045773
InChI Key VKALYYFVKBXHTF-UHFFFAOYSA-N
Synonym 3-methyl-4-methylthio phenol, 4-methylthio-m-cresol, methylthiomethylphenol, mmtp, phenol, 3-methyl-4-methylthio, 3-methyl-4-1-methylthio phenol, usaf ma-17, 4-methylthio-3-methylphenol, m-cresol, 4-methylthio, 3-methyl-4-methylsulfanyl phenol
PubChem CID 18391
ChEBI CHEBI:38681
IUPAC Name 3-methyl-4-methylsulfanylphenol
SMILES CC1=C(C=CC(=C1)O)SC

Specifications

Melting Point 54°C to 58°C
Quantity 5 g
Beilstein 1210322
Formula Weight 154.23
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)-m-cresol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.