missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Methylumbelliferyl acetate, 99%, Thermo Scientific™

Product Code. 11420123
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11420123 25 g 25g
11410123 5 g 5g
11430123 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420123 Supplier Alfa Aesar™ Supplier No. A12147.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-9-2747
ChEBI CHEBI:17763

Specifications

Melting Point 147°C to 152°C
Molecular Formula C12H10O4
MDL Number MFCD00006865
Quantity 25 g
Beilstein 189667
Synonym 7-acetoxy-4-methylcoumarin, 4-methylumbelliferyl acetate, 4-methyl-2-oxo-2h-chromen-7-yl acetate, mu-ac, 2h-1-benzopyran-2-one, 7-acetyloxy-4-methyl, 7-acetyl-4-methylcoumarin, beta-methylumbelliferyl acetate, 7-acetyloxy-4-methyl-2-benzopyrone, 4-methyl-2-oxochromen-7-yl acetate, 4-methyl-7-acetyloxy coumarin
InChI Key HXVZGASCDAGAPS-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC(=O)C
IUPAC Name (4-methyl-2-oxochromen-7-yl) acetate
Molecular Weight (g/mol) 218.21
PubChem CID 366
Formula Weight 218.21
Percent Purity 99%
Chemical Name or Material 4-Methylumbelliferyl acetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.