missing translation for 'onlineSavingsMsg'
Learn More

4-Methylvaleric Acid Methyl Ester, TRC

Product Code. 30464842
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464842 5 g 5g
30483188 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464842 Supplier Toronto Research Chemicals Supplier No. TRCM3385055G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2412-80-8
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.18
Synonym 4-Methylpentanoic Acid Methyl Ester, Methyl 4-Methylpentanoate, Methyl 4-Methylvalerate, Methyl Isocaproate, Methyl Isohexanoate
IUPAC Name methyl 4-methylpentanoate
SMILES COC(=O)CCC(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H14O2/c1-6(2)4-5-7(8)9-3/h6H,4-5H2,1-3H3
Formula Weight 130.0994
Chemical Name or Material 4-Methylvaleric Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.