missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholine-4-sulfonyl)-benzenesulfonyl chloride, Technical Grade, Thermo Scientific™

Product Code. 10643793
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10643793 1 g Amber Glass Bottle 1g
10570922 250 mg Amber Glass Bottle 250mg
10071624 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10643793 Supplier Thermo Scientific Maybridge Supplier No. MO00516DA
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-13-0
Molecular Formula C10H12ClNO5S2
Molecular Weight (g/mol) 325.778
InChI Key ZZSBUDUHESWZJB-UHFFFAOYSA-N
Synonym 4-morpholin-4-ylsulfonyl benzenesulfonyl chloride, 4-morpholine-4-sulfonyl benzene-1-sulfonyl chloride, 4-morpholinosulfonyl benzene-1-sulfonyl chloride, 4-morpholine-4-sulfonyl benzenesulfonyl chloride, 4-morpholine-4-sulfonyl-benzenesulfonyl chloride, 4-morpholine-4-sulfonyl phenylsulfonyl chloride, 4-morpholine-4-sulfonyl-benzenesulfonylchloride
PubChem CID 2779790
IUPAC Name 4-morpholin-4-ylsulfonylbenzenesulfonyl chloride
SMILES C1COCCN1S(=O)(=O)C2=CC=C(C=C2)S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 325.79
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(Morpholine-4-sulfonyl)-benzenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.