missing translation for 'onlineSavingsMsg'
Learn More

4-(Morpholinomethyl)benzoic acid, Thermo Scientific™

Product Code. 10245474
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10245474 10 g Amber Glass Bottle 10g
10127892 250 mg Amber Glass Bottle 250mg
10012172 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245474 Supplier Thermo Scientific Maybridge Supplier No. CC29301EA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 62642-62-0
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.256
InChI Key QYBXZYYECZFQRX-UHFFFAOYSA-N
Synonym 4-morpholinomethyl benzoic acid, 4-morpholin-4-ylmethyl benzoic acid, 4-morpholin-4-ylmethyl-benzoic acid, 4-4-morpholinemethyl benzoic acid, 4-4-morpholinylmethyl benzoic acid, benzoic acid, 4-4-morpholinylmethyl, timtec-bb sbb011465, cbmicro_000050, acmc-1b2v0, 4-morpholinomethylbenzoic acid
PubChem CID 703507
IUPAC Name 4-(morpholin-4-ylmethyl)benzoic acid
SMILES C1COCCN1CC2=CC=C(C=C2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 221.25
Percent Purity 97%
Chemical Name or Material 4-(Morpholinomethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.